Documenti
Z707724
TPP® centrifuge tubes
capacity 15 mL, polypropylene
Sinonimo/i:
TPP Centrifuge Tubes, TPP Tubes
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
Materiali
cap
conical bottom tube
polypropylene
transparent tube
Sterilità
sterile
Caratteristiche
RCF 15,500 × g
Confezionamento
pack of 800 ea
Produttore/marchio commerciale
TPP 91015
Capacità
15 mL
Descrizione generale
The TPP 50 mL and 15 mL Centrifuge Tubes are compatible with all standard centrifuges and their innovative design focuses on a tight seal, excellent transparency, clear graduations, and highly visible marking areas. The yellow screw cap closure on the TPP Centrifuge Tubes is tight and protected against overturning. The transparency of the tubes is excellent and the white marking areas on the top of the cap and side of the tube are highly visible. On the conical tubes, the distinct white graduations are continued to the bottom of the tube to accommodate small sample volumes. On the 500 mL tubes, the scale starts at 0.5 mL, and on the 15 mL tubes, the scale starts at 0.1mL.
Temperature Range for Polypropylene Tubes: -196°C to +121°C; Polystyrene Tubes: -40°C to +70°C; Maximum RCF for Polypropylene Tubes: 15500 × g*; Polystyrene Tubes: 1700 × g*.
* Depending on adapter and temperature
All Tubes are gamma Irradiated
Note legali
TPP is a registered trademark of TPP Techno Plastic Products AG
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
BRAND® centrifuge tubes capacity 10 mL, polypropylene, without cap, pack of 250 ea
Z640883
BRAND® centrifuge tubes