Documenti
Z333980
BRAND® pipette fillers
Standard, for pipettes up to 10 mL
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Materiali
natural rubber
Descrizione
3 valves
Caratteristiche
Standard
for pipettes up to 10 mL
Produttore/marchio commerciale
BRAND 25300
Volume
10 mL
Descrizione generale
Simple pipetting aids for volumetric and graduated pipettes. Control of the functions by squeezing the appropriate valves between thumb and fore finger.
Altre note
BR25300 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Safety bulb® pkg of 1 ea
Z118923
Safety bulb®
Pipette bulb volume 3 mL
Sigma-Aldrich
Z509043
Pipette bulb