Documenti fondamentali
Z327786
BRAND® graduated pipette, PP, high clarity
capacity 0-2 mL , 0.020 accuracy
Sinonimo/i:
graduated pipet
About This Item
Codice UNSPSC:
41121500
Materiali
polypropylene
Descrizione
blue ring mark
Caratteristiche
graduations
subdivision 0.1 mL
Confezionamento
pack of 12
Produttore/marchio commerciale
BRAND 27610
Capacità
0-2 mL , 0.020 accuracy
D.E. tubo aspirazione
≤8 mm
Descrizione generale
Resistant to breakage. Ideal for applications where glass pipettes cannot be used, e.g. in fluoro and silicate chemistry. Exposure to temperatures up to 60°C will not cause permanent exceeding of error limits. Cleaning temperatures below 60°C is recommended to preserve marks and inscriptions
Altre note
BR27610 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
This page contains detailed information on working with and handling pipettes and pipetting aids.