Documenti fondamentali
Z324876
Sigma-Aldrich
BRAND® SILBERBRAND ETERNA volumetric pipettes, calibrated to deliver
capacity 25 mL
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Materiali
AR-Glas®
agenzia
according to DIN 12 690
Caratteristiche
class B (callibrated to deliver)
class B
one-mark
Confezionamento
pack of 6 ea
Produttore/marchio commerciale
BRAND 29515
Campo di misura del volume
25 mL, accuracy: 0.04 mL
Capacità
25 mL
Descrizione generale
The BRAND® SILBERBRAND ETERNA volumetric pipettes are expertly crafted fromAR-Glas® and conform to DIN EN ISO 648 standards, ensuring high precision and reliability. Calibrated to deliver (TD, Ex), these pipettes feature a waiting time of 5 seconds for optimal accuracy in liquid transfer. Each BLAUBRAND® bulb pipette is supplied with a batch number and includes one batch certificate per packing unit from the manufacturer, guaranteeing traceability and quality. This combination of quality materials and thorough documentation makes these pipettes an excellent choice for demanding laboratory environments.
Altre note
BR29515 (India)
Note legali
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
This page contains detailed information on working with and handling pipettes and pipetting aids.