Documenti fondamentali
TMO312006-9025
Nalgene® diagnostic bottle
natural polypropylene copolymer, volume 8 mL, case of 2000 ea
Sinonimo/i:
PPCO bottles, small volume bottle with closure
Scegli un formato
2000 EACH
CHF 1’610.00
CHF 1’610.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
2000 EACH
CHF 1’610.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 1’610.00
Check Cart for Availability
Materiali
natural polypropylene copolymer
Confezionamento
case of 2000 ea
Produttore/marchio commerciale
Nalgene 312006-9025
Capacità
8 mL
Volume
8 mL
Descrizione generale
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)