Documenti fondamentali
TMO2401-0125
Nalgene® economy wash bottle
low-density polyethylene, volume 125 mL, unlabelled, case of 48 ea
Sinonimo/i:
LDPE flush bottle, LDPE wash bottle
Scegli un formato
48 EACH
CHF 289.00
CHF 289.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
48 EACH
CHF 289.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 289.00
Check Cart for Availability
Materiali
low-density polyethylene
Confezionamento
case of 48 ea
Produttore/marchio commerciale
Nalgene 2401-0125
Capacità
125 mL
Volume
125 mL
Descrizione generale
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)