Documenti fondamentali
TMO235307
Nunc® low fluorescence sealing tape
polyolefin silicone, non-sterile, case of 100 ea
Sinonimo/i:
adhesive film, adhesive tape
Scegli un formato
100 EACH
CHF 556.00
CHF 556.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
100 EACH
CHF 556.00
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.15
CHF 556.00
Check Cart for Availability
Sterilità
non-sterile
Confezionamento
case of 100 ea
Produttore/marchio commerciale
Nunc 235307
Descrizione generale
“Nunc”Sealing Tapes are effective at reducing evaporation and shielding samples from contamination or accidental spills. These adhesive seals are compatible with various assay applications. Nunc Life Science products guarantee dependable and consistent results, adhering to the highest quality standards and certifications that are reliable within the industry.
Caratteristiche e vantaggi
  • Adhesive seals are highly efficient in sealing all microplate formats.
  • Pressure sensitive adhesive is not tacky to the touch but creates a solid bond with plates when pressure is applied.
  • Thin, lightweight tapes can be applied directly onto the plate using a handheld applicator.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)