OH30073986
Ohaus® Pen pH Meter
Model ST20D
About This Item
Codice UNSPSC:
41115603
Prodotti consigliati
Materiali
epoxy shaft
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30073986
dissolved oxygen measuring range
0.0-19.9 mg/L
Campo di misura della temperatura
0.0-99.0 °C
Batteria
4 × 1.5 V, 200 hr life
Descrizione generale
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Applicazioni
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Caratteristiche e vantaggi
Easily replaceable electrodes, automatic shut off feature
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)