Vai a
MTCR8014
MTC™ Bio IceRack™ Tube Platform with Detachable Lid
to hold, 100 x 1.5 mL tubes
Sinonimo/i:
cooled microtube rack, cooled tube rack, benchtop cold tube holder, cooled tube holder, MTC Bio T8014
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
NACRES:
NB.12
UNSPSC Code:
41122806
Manufacturer/tradename:
MTC™ Bio R8014
Feature:
to hold 100 x 1.5 mL tubes
Material:
polypropylene
Packaging:
pack of 6 ea
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
polypropylene
feature
to hold 100 x 1.5 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
MTC™ Bio R8014
parameter
-80-135 °C temp. range
General description
IceRack™ boxes have a removable white tube tray that allows the blue base to be filled with crushed ice or other temperature-controlling materials. They also feature a translucent cover. Temperature-sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. These boxes are suitable for use with screw-cap or snap-cap microtubes from 0.5mL to 1.5/2.0mL.
IceRack™ for 100 × 1.5mL tubes, detachable lid, tube platform, and base, numbered cavities, accepts snap cap and screw cap 1.5mL to 2.0mL tubes, 6/cs
Features and Benefits
  • Designed for crushed Ice
  • Polypropylene, -80°C to 135°C
  • Detachable “lift-off” lids
  • Economical
  • For screw cap and snap cap tubes
Legal Information
IceRack is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
MTCR8013MTCR8012MTCR1040B
description
to hold, 100 x 1.5 mL tubes
description
to hold, 72 x 1.5 mL tubes
description
to hold, 72 x 0.5 mL tubes
description
holds 80 x 1.5 /2.0 mL tubes, blue, pack of 5 ea
feature
to hold 100 x 1.5 mL tubes
feature
to hold 72 x 1.5 mL tubes
feature
to hold 72 x 0.5 mL tubes
feature
holds 80 x 1.5 /2.0 mL tubes
manufacturer/tradename
MTC™ Bio R8014
manufacturer/tradename
MTC™ Bio R8013
manufacturer/tradename
MTC™ Bio R8012
manufacturer/tradename
MTC™ Bio R1040-B
material
polypropylene
material
polypropylene
material
polypropylene
material
blue , polypropylene
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 5 ea
parameter
-80-135 °C temp. range
parameter
-80-135 °C temp. range
parameter
-80-135 °C temp. range
parameter
-
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)