Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|
Informazioni su questo articolo
material
cardboard
feature
Holds 20 x slides
manufacturer/tradename
Heathrow Scientific HS9914
size
208x341x9 mm
color
green, trim
Features and Benefits
- Easily organize slides with the index on the front cover that corresponds with the number grid inside
- Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
- Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
- Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
- Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
- Disposable
1 of 1
Questo articolo | |||
|---|---|---|---|
| description Holds 20 x slides, green, trim | description Holds 20 x slides 76x26 mm, black (trim) | description Holds 20 x slides 76x26 mm, red (trim) | description Box only (without partitions), Holds 15 and 50 mL tubes, pk of 10 |
| feature Holds 20 x slides | feature Holds 20 x slides 76x26 mm | feature Holds 20 x slides 76x26 mm | feature Holds 15 and 50 mL tubes |
| manufacturer/tradename Heathrow Scientific HS9914 | manufacturer/tradename Heathrow Scientific HS9902 | manufacturer/tradename Heathrow Scientific HS120196 | manufacturer/tradename Heathrow Scientific HS2860M |
| material cardboard | material black (trim) | material red (trim), cardboard | material cardboard |
| size 208x341x9 mm | size - | size - | size 145x145x122 mm |
| color green, trim | color - | color - | color - |
Still not finding the right product?
Explore all of our products under Cardboard Slide Tray
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?