Documenti fondamentali
HS120647
Reagent Reservoir
capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Sinonimo/i:
Reagent container, Reagent storage
Scegli un formato
200 EACH
CHF 231.00
CHF 231.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
200 EACH
CHF 231.00
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.01
CHF 231.00
Check Cart for Availability
Materiali
V-bottom bottom
polystyrene
Sterilità
sterile; electron beam irradiated
Confezionamento
pack of 200 (packed as 5 ea)
Produttore/marchio commerciale
Heathrow Scientific 120647
Capacità
100 mL
Descrizione generale
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Caratteristiche e vantaggi
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)