Documenti fondamentali
EP5895118003
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x1000, non aerosol tight, to hold, 8 x MTP/DWP plates/tubes, with 4 high-capacity 1000 mL buckets
Scegli un formato
1 EA
CHF 4’600.00
CHF 4’600.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 4’600.00
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
CHF 4’600.00
Check Cart for Availability
Caratteristiche
to hold 8 x MTP/DWP plates/tubes
non aerosol tight
swing-bucket rotor
with 4 high-capacity 1000 mL buckets
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5895118003 (Global)
Parametri
3,153 × g max. RCF
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5920R
Descrizione generale
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)