Documenti
EP5702747009
Eppendorf® Fixed-angle rotor
F-45-18-17 Cryo, without lid
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
Caratteristiche
to hold 18 x tubes with screw cap (max o.d. 17 mm)
F-45-18-17 Cryo, without lid
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5702747009
Parametri
4,400 max. RCF (1,970 xg)
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5702/5702R/5702RH
Note legali
Eppendorf is a registered trademark of Eppendorf AG