Documenti fondamentali
EP4960800139
Eppendorf® Bottle Thread Adapter
from GL 32 to GL 27, polypropylene
Scegli un formato
1 EA
CHF 12.70
CHF 12.70
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 12.70
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.17
CHF 12.70
Check Cart for Availability
Materiali
polypropylene
Caratteristiche
from GL 32 to GL 27
Produttore/marchio commerciale
Eppendorf® 4960800139
Descrizione generale
Eppendorf dispensers are designed for bottle threads of 45 mm. For other bottle threads a thread adapter can be used. The diameter of the thread adapter is imprinted on the adapter. If the bottle thread is smaller than 32 mm, a second adapter must be used. If you need a thread adapter with a higer resistance to chemicals, use a PTFE/ETFE adapter.
Applicazioni
Connection of Eppendorf dispensers to bottles with neck threads different to 45 mm.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)