Vai a
B8532
Nalgene® narrow mouth PPCO bottle
polypropylene copolymer, capacity 500 mL, translucent, with cap
Sinonimo/i:
nalgene bottle, nalgene bottles, Nalgene 2006-0016, laboratory container, receptical, vessel
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
12 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 62.90
CHF 53.46
Informazioni su questo articolo
NACRES:
NB.17
UNSPSC Code:
41121800
CHF 53.46
Prezzo di listinoCHF 62.90Risparmia il 15%
Promozione valida solo online
Per conoscere la disponibilità, visualizza il carrello
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
polypropylene copolymer bottle (PPCO), round bottle, translucent , polypropylene copolymer
joint
Joints threaded neck (28)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2006-0016
bottle diam. × H
73 mm × 170 mm
capacity
500 mL
General description
For lab applications requiring excellent chemical resistance and autoclaving, with and without contents, use Nalgene®arrow-mouth PPCO bottles with Closure. These polypropylene copolymer bottles are translucent, providing good clarity for viewing contents. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features and Benefits
  • Nalgene® narrow-mouth bottles are ideal for long-term storage.
  • Made from polypropylene copolymer (PPCO) material for excellent sample clarity.
  • Highly chemical resistant to most acids, bases, and alcohols.
  • Autoclavable - with and without contents.
  • Laboratory-grade plastic materials for dependably low leachables and extractables.
  • Liner less caps ensures leak-proof seal for sample protection.
  • Light-weight - enables easy lifting and transporting.
  • Avoid UV exposure.
  • Manufactured from non-toxic materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
B8282B8407B8157
description
polypropylene copolymer, capacity 500 mL, translucent, with cap
description
polypropylene copolymer, capacity 125 mL, translucent, with cap
description
polypropylene copolymer, capacity 250 mL, translucent, with cap
description
polypropylene copolymer, capacity 60 mL, translucent, with cap
manufacturer/tradename
Nalgene® 2006-0016
manufacturer/tradename
Nalgene® 2006-0004
manufacturer/tradename
Nalgene® 2006-0008
manufacturer/tradename
Nalgene® 2006-0002
feature
with cap
feature
with cap
feature
with cap
feature
with cap
material
round bottle, polypropylene copolymer bottle (PPCO), translucent
material
translucent , polypropylene copolymer bottle (PPCO), round bottle
material
round bottle, polypropylene copolymer bottle (PPCO), translucent
material
polypropylene copolymer , round translucent bottle, polypropylene copolymer bottle (PPCO)
capacity
500 mL
capacity
125 mL
capacity
250 mL
capacity
60 mL
joint
Joints threaded neck (28)
joint
Joints threaded neck (24)
joint
Joints threaded neck (24)
joint
Joints threaded neck (20)
bottle diam. × H
73 mm × 170 mm
bottle diam. × H
50 mm × 101 mm
bottle diam. × H
61 mm × 133 mm
bottle diam. × H
37 mm × 85 mm
Still not finding the right product?
Explore all of our products under Nalgene® narrow mouth PPCO bottle
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)