Documenti fondamentali
EP0030119860
Eppendorf® Single Tube Stand for Conical Tubes 25 mL and 50 mL
case of 4 pieces, white, autoclavable
Scegli un formato
1 PKG
CHF 26.40
CHF 26.40
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 PKG
CHF 26.40
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
CHF 26.40
Check Cart for Availability
Materiali
white
Caratteristiche
autoclavable
Confezionamento
case of 1 ea
case of 4 pieces
Produttore/marchio commerciale
Eppendorf® 0030119860
Categorie correlate
Descrizione generale
  • Stackable within one rack format and also between all rack variants?–?saves laboratory space when not in use
  • Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
  • Wavelike design: Tube rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable
  • Made of polypropylene (PP) for high chemical and physical resistance
  • Optimized space-saving dimensions due to perfect tube position-to-footprint arrangement without compromising the tube handling
  • High flexibility: Different sized tubes fit in the 1.5/2.0 mL, 5.0/15.0 mL, 5.0/15/50 mL, and cryogenic tube rack
  • Numbered tube positions support fast and easy orientation and error-free handling
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)