Documenti
EP0030076265
Eppendorf® Empty Box 2.0 for epT.I.P.S.® Reloads
new
for tips 1250 µL – 5 mL
About This Item
Codice UNSPSC:
41121600
Caratteristiche
for tips 1250 µL – 5 mL
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Eppendorf® 0030076265
Descrizione generale
epT.I.P.S.® Box 2.0 for epT.I.P.S.® Reloads, empty box, does not include tips, Eppendorf Quality, 1,250 µL L – 5 mL
Applicazioni
General liquid handling
Caratteristiche e vantaggi
  • epT.I.P.S.® Box 2.0 – empty reusable boxes for tip volumes of 10 µL - 5 mL
  • epT.I.P.S.® Box 2.0 – all three sizes are availabe in Eppendorf Quality
  • Autoclavable at 121 °C (20 min.) for subsequent use
Note legali
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG