Documenti fondamentali
CLS432078
Corning® CoolCell FTS30
removable cryogenic vial module
Sinonimo/i:
Corning Coolcell accessory
Scegli un formato
10 EA
CHF 143.00
CHF 143.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 143.00
About This Item
Codice UNSPSC:
41103025
NACRES:
NB.12
CHF 143.00
Check Cart for Availability
Confezionamento
pkg of 10
Produttore/marchio commerciale
Corning 432078
Categorie correlate
Descrizione generale
CoolCelll FTS30 utilizes a solid state core and controlled microconvection technology to evenly draw in -80°C freezer air through a bottom base vent, uniformly disperse the cold air around each vial in the central chamber and then release the thermal load from the vials through a top vent choke. The inner vial module holds 30 cryogenic vials and can be removed in one step. Each vial achieves a uniform and reproducible -1°C/minute freezing profile and thermal profiles are highly reproducible. Due to the low thermal mass of the uniform-density cross-linked polyethylene foam container, freezing can be conducted without a rise in local freezer temperature, thereby protecting nearby samples.
  • For 30 standard 1.0 mL to 2.0 mL cryogenic vials.
  • Controlled micro-convection for uniform freezing of 30 vials.
  • Removable vial tray for one-step transfer of samples into and out of freezing chamber.
Note legali
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)