Documenti fondamentali
CLS431227
Corning® syringe filters
PTFE membrane, diam. 50 mm, pore size 0.2 μm
Sinonimo/i:
disposable syringe filters, sterile syringe filters, syringe filters
Scegli un formato
12 EA
CHF 229.00
CHF 229.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EA
CHF 229.00
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
CHF 229.00
Check Cart for Availability
Materiali
PTFE membrane
polypropylene housing
Sterilità
sterile
Confezionamento
case of 12
Produttore/marchio commerciale
Corning 431227
Diametro
50 mm
Dimensione pori
0.2 μm
Raccordi
hose barb
Descrizione generale
Corning syringe filters are 100% integrity tested and are certified nonpyrogenic and noncytotoxic and are manufacturered in accordance with ISO 9002 standards. A variety of membranes are available to meet your needs.
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
These Corning syringe filters have polypropylene copolymer housings, hose barb inlets and hose barb outlets.
  • PTFE membranes provide the best chemical resistance
  • These naturally hydrophobic membranes can be used for venting applications or filtering gases
  • 100% integrity tested, sterile and certified nonpyrogenic and noncytotoxic
  • Manufactured in accordance with ISO 9002 standards
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)