CLS3961
Corning® 96 well Polypropylene Deep Well Plate
V-bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no
Sinonimo/i:
cell culture plate, multi well plates, multiple well plates
Scegli un formato
CHF 1’320.00
Scegli un formato
About This Item
CHF 1’320.00
Prodotti consigliati
Materiali
V-bottom clear
polypropylene
polypropylene plate
translucent bottom
Sterilità
non-sterile
Caratteristiche
lid: no
skirt
Produttore/marchio commerciale
Corning 3961
Confezionamento
bag of 5 ×
Misura
96 wells
Volume pozzetto
2 mL
Pozzetti
96 wells , storage block
Compatibilità
suitable for (cell cuture)
Tipo di binding
non-treated surface
Categorie correlate
Descrizione generale
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)