Documenti fondamentali
CLS2975225
Corning® cover glasses
rectangular, No. 1, W × L 22 mm × 50 mm
Sinonimo/i:
coverslips, glass covers, slide covers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.16
Materiali
borosilicate glass
rectangular
Confezionamento
pack of 100 ea
case of 1000 ea
Produttore/marchio commerciale
Corning 2975-225
Largh. × lungh.
22 mm × 50 mm
Tipo di binding
non-treated surface
Caratteristiche e vantaggi
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
BRAND® cover glass L × W 24 mm × 60 mm, rectangular
BR470820
BRAND® cover glass
Cover glasses size 24 mm × 60 mm
Sigma-Aldrich
C9056
Cover glasses
Corning® cover glasses square, No. 11/2, W × L 22 mm × 22 mm
CLS285022
Corning® cover glasses
Corning® cover glasses square, No. 1, W x L 18 mm x 18 mm
CLS284518
Corning® cover glasses
Cover glasses size 22 mm × 22 mm
C9802
Cover glasses
Corning® cover glasses rectangular, No. 11/2, W × L 24 mm × 60 mm
CLS2980246
Corning® cover glasses