Documenti
CLS285025
Corning® cover glasses
square, No. 11/2, W × L 25 mm × 25 mm
Sinonimo/i:
coverslips, glass covers, slide covers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.16
Materiali
borosilicate glass
square
Confezionamento
pack of 200 ea
case of 2000 ea
Produttore/marchio commerciale
Corning 2850-25
Largh. × lungh.
25 mm × 25 mm
Tipo di binding
non-treated surface
Caratteristiche e vantaggi
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Note legali
Corning is a registered trademark of Corning, Inc.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Cover glasses size 22 mm × 22 mm
C9802
Cover glasses