CLS11651
Corning® closed system bottle
capacity 150 mL, polystyrene, male MPC connector, sterile
Sinonimo/i:
storage vessel
Scegli un formato
CHF 1’950.00
Scegli un formato
About This Item
CHF 1’950.00
Prodotti consigliati
Materiali
clear bottle
high-density polyethylene cap
polystyrene
round bottle
wide-mouth bottle
Sterilità
sterile
Caratteristiche
cap
male MPC connector
Confezionamento
pkg of 8 ea
Produttore/marchio commerciale
Corning 11651
Diam. del collo della bottiglia
~42 mm
Capacità
150 mL
Categorie correlate
Descrizione generale
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
- Made from optically clear polystyrene
- Sterilized to SAL 10-6
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)