Documenti fondamentali
BMSW1005500
Sigma-Aldrich
Accuris M1 Grade Calibration Weight
size 500 g, stainless steel
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.71
Materiali
stainless steel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Benchmark Scientific W1005-500
Misura
500 g
Descrizione generale
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Note legali
Accuris is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)