Documenti fondamentali
BAF134730000
Bel-Art® Write-on Tape Dispenser
Economy model, Multi-Roll
Sinonimo/i:
label tape dispenser, tape cutter
Scegli un formato
1 EA
CHF 62.80
CHF 62.80
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 62.80
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
CHF 62.80
Check Cart for Availability
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F13473-0000
Lungh. × Largh. × Alt.
30.2 cm × 14.3 cm × 12.1 cm
Descrizione generale
Provides a smooth writing surface, making labeling easy and fast. When marked tape is advanced and cut off on the serrated edge, a fresh length of label tape is positioned on the writing platform. Plastic cutting edge is molded into the housing. Various width and color tape rolls can be mixed together on the spindles, which accommodates 5 or more 12 – 19mm ( ½ – ¾”) narrow rolls or 4 – 25mm (1”) wide rolls. Spindles are easily removed from the unit for loading and unloading, and can be adjusted to hold either 25 or 75mm (1 or 3”) core diameter rolls.
Caratteristiche e vantaggi
Includes three plastic separating discs to assure smooth, snag free dispensing for up to four rolls of tape; Additional separating discs are available separately.
Molded-in pen holder wells on the dispenser help to keep writing instruments handy
Ideal for use with the full line of 13463-series Write-On Tapes, which are available in 6 colors and 3 widths; Includes one free roll of ½” White, Write-On Labeling Tape.
Includes mounting holes for secure attachment to a benchtop (screws not included); Can also be attached to vertical surfaces using Wall Mount Adapter (not included, BAF134730001).
Altre note
The Economy Write-On Tape Dispenser is intended for simpler uses, as the plastic cutting edge is molded into the housing. Mounting holes in the base of the unit can be used to attach to the bench top.
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)