Documenti fondamentali
B5910
Sigma-Aldrich
Nalgene® bottles, style 2007
capacity 500 mL
Sinonimo/i:
HDPE bottles, nalgene bottle, nalgene bottles
Scegli un formato
12 EA
CHF 87.10
CHF 87.10
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EA
CHF 87.10
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 87.10
Check Cart for Availability
Materiali
high-density polyethylene bottle
polypropylene cap
polypropylene screw closure
rectangular translucent bottle
Giunto
Joints threaded neck (48-415)
Descrizione
Not autoclavable
Caratteristiche
with cap
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Nalgene 2007-0016
Lungh. &Times; Alt. &Times; Prof. bottiglia
94 mm × 117 mm × 58.7 mm
Capacità
500 mL
Descrizione generale
Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. and have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)