Documenti fondamentali
ZTLP0EDI1
Milli-Q
Modulo di elettrodeionizzazione (EDI) Elix®
Water Purification by Electrodeionization (EDI)
About This Item
Codice UNSPSC:
41104200
NACRES:
JA.13
Prezzi e disponibilità al momento non sono disponibili
Confezionamento
pkg of 1 unit
Produttore/marchio commerciale
Elix®
Altezza dell’unità
33 cm (13 in.)
Descrizione generale
The Elix® EDI module is designed to continuously deionize the water, post-reverse osmosis by electrodeionization technique. The Elix® EDI module delivers superior quality pure water consistently and reliably, without any maintenance.
Applicazioni
The Type 2 water produced post RO-EDI treatment meets the needs of many general laboratory applications, and for use in clinical analyzers, cell culture incubators and weatherometers.
Caratteristiche e vantaggi
  • Efficiently removes ions.
  • High water recovery.
  • Low maintenance.
Note legali
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany