Documenti fondamentali
SPKGENPNE001
Millipore
Spare part, Filter Cartridge
40µm Filtration grade
About This Item
Codice UNSPSC:
23151800
Prezzi e disponibilità al momento non sono disponibili
Materiali
plastic
Livello qualitativo
Nome Commerciale
Mobius®
Compatibilità
for use with Mobius®
Descrizione generale
This cartridge filter is designed to remove contaminants such as water and particles from compressed air systems. Therefore, it ensures the compressed air quality.
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.
Confezionamento
Pack of 1 unit
Note legali
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany