Documenti fondamentali
2402-M
Millipore
Stat-Matic II Plate Washer, with bottle
The StatMatic II automated plate washer is designed for use with 96-well microplates. Internal volume calibration rings allow you to adjust volumes from 100, 200, 250 & 300uL per well from a single-dispense stroke.
Sinonimo/i:
plate washer
About This Item
Codice UNSPSC:
41104923
eCl@ss:
36102014
Produttore/marchio commerciale
Chemicon®
Descrizione generale
Ideal for hand-held applications or for use in an automated line. The StatMatic II with a hand-held eight-port disposable manifold provides greater ease, speed, convenience, and affordability than a multi-channel pipette. The StatMatic II can be connected to a range of bottle sizes and can completely fill a microplate with one-handed operation -- in a matter of seconds.
The StatMatic II is designed for use with 96-well microplates. Internal volume calibration rings allow you to adjust volumes from 100, 200, 250, and 300uL per well from a single-dispense stroke.
The PTFE plunger guarantees smooth and accurate dispensing every time. Compatible with most reagents and chemicals, the design ensures that even protein-based reagents won′t cause sticking.
Note legali
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Esclusione di responsabilità
General Lab Use
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4