Documenti fondamentali
Z743209
Julabo® bath accessories
hollow polyproplylene balls, diam. 20 mm, for use in Julabo CORIO and Pura water baths, pack of 1000 pieces
Scegli un formato
1 EA
CHF 149.00
CHF 149.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 149.00
About This Item
Codice UNSPSC:
41103706
NACRES:
NB.77
CHF 149.00
Check Cart for Availability
Materiali
polypropylene
Confezionamento
pack of 1000 pieces
Produttore/marchio commerciale
Julabo 8970010
Descrizione generale
Place the hollow polyproplylene balls in a thermostat bath to form an even covering layer, exclusively for use in waterr. The floating balls reduce: heat loss, evaporation loss (up to 90%), oxygen input, odors, light exposure and are food-safe up to 100°C.
Note legali
Julabo is a registered trademark of Julabo USA, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)