Documenti fondamentali
Z743206
Julabo® bath accessories
assembly cooling coil, for CORIO C and CD ranges, pack of 1 ea
Scegli un formato
1 EA
CHF 131.00
CHF 131.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 131.00
About This Item
Codice UNSPSC:
41103706
NACRES:
NB.77
CHF 131.00
Check Cart for Availability
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Julabo 9970100
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 4
Questo articolo
Z743214Z743212Z743211
vibrant-m
Z743206
Julabo® bath accessories
Julabo® bath accessories installation cooing coil, for CORIO C and CD ranges, pack of 1 ea
Z743214
Julabo® bath accessories
Julabo® bath accessories immersion-height adjustable platform, for CORIO C-B19/27 and CD-B19/27, pack of 1 ea
Z743212
Julabo® bath accessories
Julabo® bath accessories immersion-height adjustable platform, for CORIO C-B13/17 and CD-B13/17, pack of 1 ea
Z743211
Julabo® bath accessories
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
Descrizione generale
Built-in cooling coil (9mm outer diameter) for counter-cooling with cooling water, can be used for tempering tasks close to the ambient temperature. A hose with an inner diameter of 10/12 mm is required. For use with Julabo® baths/circulators.
Note legali
Julabo is a registered trademark of Julabo USA, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)