Documenti fondamentali
Z742514
Block for digital dry bath and MyBlock
solid block for slides or custom machining
About This Item
Codice UNSPSC:
41102404
NACRES:
NB.77
Caratteristiche
solid block for slides or custom machining
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Benchmark Scientific BSW01
Descrizione generale
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Note legali
MyBlock is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)