Documenti
Z742501
MyBlock mini dry bath block
to hold, 15 x 1.5 mL vials
About This Item
Codice UNSPSC:
41102404
NACRES:
NC.21
Caratteristiche
to hold 15 x 1.5 mL vials
for Mini dry bath with Heat Lid
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Benchmark Scientific BSH100-15HL
Descrizione generale
To fit MyBlock mini dry bath/block heater.
Note legali
MyBlock is a trademark of Benchmark Scientific, Inc.