Documenti fondamentali
Z679933
Bio-Pure pipetting reservoir
capacity 25 mL, with divider
Sinonimo/i:
multichannel pipette reservoir, reagent reservoir
Scegli un formato
100 EA
CHF 111.00
CHF 111.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
100 EA
CHF 111.00
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.01
CHF 111.00
Check Cart for Availability
Materiali
PET (Bio-degradable)
V-bottom bottom
white
Sterilità
sterile
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Diversified Biotech REBP-2000
Capacità
25 mL
Descrizione generale
Bio-Pure Reagent Reservoirs are made from recaputured PET that does not rely on additional fossil fuel-derived petrochemicals. In addition, reservoirs are sealed in bio-degradable bags and packed in recycled cardboard cases. As a result, Bio-Pure Reservoirs are a more eco-friendly alternative. "Trough within a trough" maximizes reagent recovery. A wide base and reinforced sidewalls avoids spills and provides stability. Gamma irradiated RNASE/DNASE free
Caratteristiche e vantaggi
  • Constructed from biodegradable PET material for environmentally conscious disposal.
  • Specialized "trough within a trough" design for optimal reagent recovery.
  • Pour-off spouts at all four corners and a wide base for improved stability and spill prevention.
  • Sterilized for safe and contamination-free usage.
Note legali
Bio-Pure is a trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)