Documenti fondamentali
Z566128
Sigma-Aldrich
Micromatic Macrovalve flushing adapter for returnable containers
allows nitrogen to be flushed through the Coupler and solvent transfer line
Scegli un formato
1 EA
CHF 164.00
CHF 164.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 164.00
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.43
CHF 164.00
Check Cart for Availability
Descrizione generale
This adapter allows nitrogen to be flushed through the Micromatic MacroValve Coupler and solvent transfer line when they are disconnected from the Pure-Pac® II containers.
Note legali
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)