Documenti fondamentali
Z562580
Sigma-Aldrich
Aldrich® corrosive gas regulator with electroless nickel-plated body and cross purge assembly
inlet CGA 660
Scegli un formato
1 EA
CHF 3’240.00
CHF 3’240.00
Spedizione prevista il23 maggio 2025
Scegli un formato
Cambia visualizzazione
1 EA
CHF 3’240.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
CHF 3’240.00
Spedizione prevista il23 maggio 2025
Materiali
316L stainless steel diaphragm (PTFE-lined)
Monel® nozzle
Descrizione
Captured vent
PTCFE seat and Viton® seals
delivery pressure measuring range
0-80 psig
Pressione in ingresso
3000 psig
CGA ingresso
660
NPTM uscita
1/4 in.
Descrizione generale
The combination of Monel® regulator components and proper purging with the cross-purge assembly makes these units virtually impervious to attack from strong acid forming gases that include hydrogen chloride, chlorine, hydrogen fluoride, hydrogen bromide, and silicon tetrafluoride.
Cross-purge assembly eliminates the escape of toxic and flammable gases when changing cylinders and permits purging system with dry, inert gas to prevent corrosion due to gas hydrolysis.
Specifications:
  • Single-stage
  • Electroless nickel-plated body
  • Stainless steel diaphragm
  • 316L Stainless steel cross-purge with color-coded 1/4 turn valves with 1/4 in. NPTM outlets
Request Technical Information Bulletin No. AL-202 for cross-purge techniques.
Altre note
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Monel is a registered trademark of Huntington Alloys Corp.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)