Documenti fondamentali
Z553654
Sigma-Aldrich
Aldrich® modified Friedrichs condenser with SafetyBarb® tubing connectors
jacket O.D. 54 mm, Overall H 300 mm
Scegli un formato
1 EA
CHF 1’320.00
CHF 1’320.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 1’320.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 1’320.00
Check Cart for Availability
Giunto
joint ST/NS24/40
Alt. complessiva
300 mm
D.E. camicia
54 mm
Categorie correlate
Descrizione generale
This glassware features removable polypropylene hose barb connections for 1/4in. I.D. tubing. The barbs eliminate accidental glassware breakage when installing or removing tubing. A silicone rubber seal ensures a liquid- and vacuum-tight connection to the glass GL-14 thread. Simply unscrew the cap to remove the hose barb.
Molded spiral condensing coolant tube fits closely within jacket to force vapors along the spiral path. The modified feed tube reduces chance of breakage.
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Applicazioni
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
Analyze omega 3 fatty acid profiles from fish oil capsules and salmon using GC columns for confirmatory analysis of lipid composition.
Chromatograms
suitable for GCsuitable for GCsuitable for GC