Documenti fondamentali
Z551880
Sigma-Aldrich
Aldrich® in-line oil bubbler with SafetyBarb® hose connectors
joint: ST/NS 14/20
Sinonimo/i:
bubblers, in-line bubblers, scientific bubblers
Scegli un formato
1 EA
CHF 209.00
CHF 209.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 209.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 209.00
Check Cart for Availability
Giunto
joint: ST/NS14/20
Capacità
10 mL
Categorie correlate
Descrizione generale
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)