Documenti fondamentali
Z549851
Sigma-Aldrich
Aldrich® jacketed three-neck flask
half jacket, capacity 12,000 mL, center joint: ST/NS 45/50
Sinonimo/i:
boiling flask, flask round, flask round bottom
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
round bottom
three-neck
three-neck flask
Giunto
center joint: ST/NS45/50
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
pack of 1
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
12,000 mL
Descrizione generale
Unique design which incorporates a flush-closing PTFE drain valve eliminating unwanted dead space.
Features:
  • Eliminates dead space for complete stirring of flask contents
  • Allows for complete draining without breaking down flask set-up
  • Liquid cooled or heated with liquid or steam
  • 24/40 side joints
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)