Documenti fondamentali
Z549347
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 250 mL
Scegli un formato
1 EA
CHF 311.00
CHF 311.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 311.00
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 311.00
Check Cart for Availability
Giunto
female joint: ST/NS29/32
male joint: ST/NS24/40
Capacità
250 mL
Dimensione pori
70-100 μm porosity
Categorie correlate
Descrizione generale
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)