Documenti fondamentali
Z513385
Sigma-Aldrich
Micro spatulas
flat rounded/tapered ends, L 7 in. (178 mm)
Scegli un formato
1 PKG
CHF 120.00
CHF 120.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 PKG
CHF 120.00
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
CHF 120.00
Check Cart for Availability
Materiali
nickel-stainless steel
Confezionamento
pkg of 12 ea
Lunghezza
7 in. (178 mm)
Descrizione generale
Nickel stainless spatula one end rounded, one end tapered. Rounded end approximately 5/16 in. (8 mm) wide x 2 in. (51 mm) long. Tapered end approximately 1/4 in. (6 mm) wide x 2 in. (51 mm) long.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Lab Spatula Flat end and spoon end, nickel-stainless steel (plated), L 225 mm
HS15909
Lab Spatula
PTFE-coated spatulas Tapered to 1/8in., L 8 1/4 in.
Z115258
PTFE-coated spatulas
Lab spoons stainless steel, L 7 1/2 in.
Sigma-Aldrich
Z511455
Lab spoons
PTFE-coated spatulas Tapered to 1/8in., L 7 1/4 in.
Z115282
PTFE-coated spatulas
Questions
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.