Documenti fondamentali
Z510793
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 19/22, thread size 13-425
Sinonimo/i:
747130-1319
Scegli un formato
1 EA
CHF 49.60
CHF 49.60
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 49.60
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 49.60
Check Cart for Availability
Misura filettatura
13-425
Giunto
female joint: ST/NS19/22
Descrizione generale
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Caratteristiche e vantaggi
  • Accessory for microscale MICROFLEX threaded kits
  • Connects Standard Taper outer ground joint to GPI thread of inner ground joint
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)