Documenti fondamentali
Z510831
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 24/40, thread size 20-400
Sinonimo/i:
747130-2024
Scegli un formato
1 EA
CHF 61.90
CHF 61.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 61.90
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 61.90
Check Cart for Availability
Misura filettatura
20-400
Giunto
female joint: ST/NS24/40
Descrizione generale
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Caratteristiche e vantaggi
  • Accessory for microscale MICROFLEX threaded kits
  • Connects Standard Taper outer ground joint to GPI thread of inner ground joint
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)