Documenti fondamentali
Z302457
Quickfit® Cone/Screw-thread Adapters
24 thread (couples 10.5 - 11.5 mm diameter), joint: ST/NS 29/32
Sinonimo/i:
ST54/24
Scegli un formato
1 EA
CHF 66.40
CHF 66.40
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 66.40
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 66.40
Check Cart for Availability
Giunto
joint: ST/NS29/32
Produttore/marchio commerciale
Quickfit ST 54/24
Disponibilità
not available in USA and Canada
not available in USA and Canada
Raccordi
24 thread (couples 10.5 - 11.5 mm diameter)
Descrizione generale
The Quickfit screw-thread joints on these cone adapters provide a versatile means of coupling thermometers, bleed tubes, electrodes, etc; with Quickfit glassware. The joints are readily adjustable for depth of immersion by slackening the screw cap, and give excellent results under vacuum.
Can be used in apparatus at temperatures up to 300°C providing excessive heat is not applied directly to the screw-thread union.
For replacement parts, see related products section of web site listing.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)