Documenti fondamentali
Z302384
Quickfit® Cone/Screw-thread Adapters
18 thread (couples 7.0 - 8.5 mm diameter), joint: ST/NS 14/23
Sinonimo/i:
ST51/18
Scegli un formato
1 EA
CHF 41.80
CHF 41.80
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 41.80
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 41.80
Check Cart for Availability
Giunto
joint: ST/NS14/23
Produttore/marchio commerciale
Quickfit ST 51/18
Disponibilità
not available in USA and Canada
not available in USA and Canada
Raccordi
18 thread (couples 7.0 - 8.5 mm diameter)
Descrizione generale
The Quickfit screw-thread joints on these cone adapters provide a versatile means of coupling thermometers, bleed tubes, electrodes, etc; with Quickfit glassware. The joints are readily adjustable for depth of immersion by slackening the screw cap, and give excellent results under vacuum.
Can be used in apparatus at temperatures up to 300°C providing excessive heat is not applied directly to the screw-thread union.
For replacement parts, see related products section of web site listing.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)