Documenti
Z281255
Laboratory brushes
Test tube, L 9 in., brush diam. × L 3/4 in. × 3 1/4 in.
About This Item
Codice UNSPSC:
47131605
NACRES:
NB.35
Confezionamento
pkg of 4 ea
Lunghezza
9 in.
brush diam. × L
3/4 in. × 3 1/4 in.
Descrizione generale
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Flask brush Black nylon
Z225509
Flask brush
Laboratory brushes Flask, L 12 in., brush diam. × L 1 3/8 in. × 3 1/2 in.
Z281344
Laboratory brushes
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Pipette, L 17 in., brush diam. × L 3/16- 3/4 in. × 7 1/2 in.
Z281328
Laboratory brushes
Laboratory brushes Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.
Z281336
Laboratory brushes