Documenti
Z275301
Sigma-Aldrich
Cannon-Fenske viscometer tube
size 100
Sinonimo/i:
Viscosity Measurement Device, Viscosity Tube
About This Item
Codice UNSPSC:
41111700
NACRES:
PB.36
Misura
100
Descrizione
constant ~ 0.015 cSt/s
Produttore/marchio commerciale
Ace Glass 798839
Campo di misura della viscosità
3-15 cSt
Categorie correlate
Descrizione generale
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.