Documenti fondamentali
Z274887
Maintenance kit for KNF Laboport® solid PTFE vacuum pump
Maintenance kit for 34 Lpm pump
Scegli un formato
1 KIT
CHF 270.00
CHF 270.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 KIT
CHF 270.00
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.79
CHF 270.00
Check Cart for Availability
Produttore/marchio commerciale
KNF
Descrizione generale
Includes replacement PTFE diaphragm, head seals and two Kalrez valves.
Kits include replacement PTFE diaphragm, head seals, and two Kalrez® valves. Note: Two kits are required when servicing both heads of vacuum pump.
Altre note
Two kits are required when servicing both heads of vacuum pump.
Note legali
Kalrez is a registered trademark of E. I. du Pont de Nemours and Company
Laboport is a registered trademark of KNF Neuberger GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)