Documenti fondamentali
Z258997
Flexible stirrer shaft
L 152.4 cm (60 in.) , overall
Scegli un formato
1 EA
CHF 365.00
CHF 365.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 365.00
About This Item
Codice UNSPSC:
41103800
NACRES:
NB.72
CHF 365.00
Check Cart for Availability
Materiali
glass
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ace Glass 8081-32
Lunghezza
152.4 cm (60 in.) , overall
Descrizione generale
Used for isolating motor from stirring shaft. Designed with a ball bearing motor coupling at one end for connection to any motor with an 8 mm (5/16 in.) diam. shaft or other power source using an 8 mm diam. round steel adapter (included).
The other end has a detachable handpiece with an 8 mm diam. round steel adapter for connection to a FLEX-GRIP® stirrer chuck (available separately). Handpiece is secured with a lab clamp. Shafts operate to 14,000 rpm in a counter-clockwise direction.
Operating and service instructions for lubrication are supplied.
Complete item includes: A flexible shaft, motor coupling for 8 mm shaft, handpiece with adapter for chuck, and a chuck wrench and chain.
Note legali
Flex-Grip is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)