Documenti fondamentali
Z244023
Sigma-Aldrich
ACE Manifold with threaded stopcocks
positions, 5
Scegli un formato
1 EA
CHF 1’200.00
CHF 1’200.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 1’200.00
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.79
CHF 1’200.00
Check Cart for Availability
Caratteristiche
positions 5
Produttore/marchio commerciale
Ace Glass 872954
Categorie correlate
Descrizione generale
Greaseless vacuum/gas manifold with 0 to 4mm high-vacuum PTFE, three-way stopcocks for takeoffs. One end of each manifold tube has a #7 Ace-Thred for easy cleaning. Ends are supplied with nylon stopper/plugs with FETFE® O-rings. Port connections are 8mm O.D. Vacuum and gas connections are 1/2in. o.d.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)